Learn More
Thymine, 97%, Thermo Scientific Chemicals
Catalog Number p-7022902
Click to view available options
Quantity:
25 g
100 g
500 g
65-71-4
C5H6N2O2
126.115
MFCD00006026
RWQNBRDOKXIBIV-UHFFFAOYSA-N
thymine, 5-methyluracil, thymin, 2,4-dihydroxy-5-methylpyrimidine, thymine anhydrate, 2,4 1h,3h-pyrimidinedione, 5-methyl, 5-methylpyrimidine-2,4 1h,3h-dione, 5-methyl-2,4 1h,3h-pyrimidinedione, thymin purine base, 5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
1135
CHEBI:17821
5-methyl-1H-pyrimidine-2,4-dione
CC1=CNC(=O)NC1=O
65-71-4
C5H6N2O2
126.115
MFCD00006026
RWQNBRDOKXIBIV-UHFFFAOYSA-N
thymine, 5-methyluracil, thymin, 2,4-dihydroxy-5-methylpyrimidine, thymine anhydrate, 2,4 1h,3h-pyrimidinedione, 5-methyl, 5-methylpyrimidine-2,4 1h,3h-dione, 5-methyl-2,4 1h,3h-pyrimidinedione, thymin purine base, 5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
1135
CHEBI:17821
5-methyl-1H-pyrimidine-2,4-dione
CC1=CNC(=O)NC1=O
Thymine is one of the four nucleobases, along with adenine, guanine and cytosine found in deoxyribonucleic acids (DNA). Thymine may be used to study chemical processes that affect DNA structure, such a radiation induced radical production leading to base cross-linking reactions and derivitizations. Thymine may be used to study the parameters of hydrogen bonding kinetics and energies with other nucleobases such as adenine. Thymine is used to develop sensitive heavy metal (mercury) detectors based on coordination chemistry and nanoparticle structures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thymine is one of the four nucleobases, along with adenine, guanine and cytosine found in deoxyribonucleic acids (DNA). Thymine may be used to study chemical processes that affect DNA structure, such a radiation induced radical production leading to base cross-linking reactions and derivitizations. Thymine may be used to study the parameters of hydrogen bonding kinetics and energies with other nucleobases such as adenine. Thymine is used to develop sensitive heavy metal (mercury) detectors based on coordination chemistry and nanoparticle structures.
Solubility
Soluble in hot water. Slightly soluble in alcohol.
Notes
Store in cool dry place.. Keep away from oxidizing agents and acids.
TRUSTED_SUSTAINABILITY
Melting Point 316°C to 317°C
Assay Percent Range 97%
Quantity 25 g
Beilstein 607626
Merck Index 14,9398
Solubility Information Soluble in hot water. Slightly soluble in alcohol.
Formula Weight 126.12
Percent Purity 97%
Chemical Name or Material Thymine