missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
BD Biosciences™ THROMBOCYTEST™ (CE/IVD)
Beskrivelse
- Contains various polyclonal antibodies labelled with the fluorescence dye phycoerythrin (PE) as follows:
- Antibodies against total human immunoglobulins, against human IgA, IgM and IgG and polyclonal antibodies against rabbit immunoglobulin as a negative control.
- The test kit also contains a fluorescence isothiocyanate (FITC) conjugated monoclonal antibody that recognizes a platelet-specific antigen and further reagents.
- Fluorescence 2 intensity values correspond to the amount of platelet associated immunoglobulin.
- Quantitation of platelet-associated immunoglobulin should be performed by flow cytometry.
- Detailed instructions result from specific experience and precise validation assays.
Tekniske data
Tekniske data
| Indhold og opbevaring | Store the kit in the dark at 2-8°C. The reagents are supplied sterile with a preservative that does not influence the performance of the assay. The reagents are stable for the period shown on the packaging label, when stored as described. |
| Inkluderer | 1 Kit: 2 x 50 mL 10X Wash Buffer, 1.25 mL Goat Serum, 0.5 mL Polyclonal Antibody to Rabbit Immunogloblulins, 0.5 mL Polyclonal Antibody to Total Human Immunoglobulins, 0.5 mL Polyclonal Antibody to Human IgA, 0.5 mL Polyclonal Antibody to Human IgG, 0.5 mL Polyclonal Antibody to Human IgM, 2.5 mL anti-CD42a-FITC |
| Antal prøver | 25 Tests |
| Mængde | 25 tests |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?