missing translation for 'onlineSavingsMsg'
Få mere at vide

Thorium Standard: Th @ 10000 μg/mL in 5% HNO3, VHG™

Artikelnummer. 30003185
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 ml
250 ml
500 ml
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003185 500 ml
30003131 250 ml
30002972 100 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30003185 Leverandør VHG Labs Leverandørnr. VHGTTHN500

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Thorium Standard: Th @ 10000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Thorium
CAS 7440-29-1
Metoder ICP/ICP-MS
Analyt- eller komponentnavne Thorium
Forsendelsestilstand Room Temperature
Molekylær formel Th
Koncentration 10000 μg/mL
Certificeringer/compliance ISO 17034
Synonym Th
InChI formel InChI=1S/Th
SMIL [Th]
IUPAC navn thorium
Opløsningsmiddel eller Matrix Dilute HNO3
Molekylvægt (g/mol) 232.038
Mængde 500 ml
Formel vægt 232.038 g/mol
Vis mere Vis mindre
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.