missing translation for 'onlineSavingsMsg'
Få mere at vide

Thorium Standard: Th @ 1000 μg/mL in 5% HNO3, VHG™

Artikelnummer. 17968769
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 ml
100 ml
250 ml
500 ml
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17968769 500 ml
17909389 250 ml
17997979 50 ml
17937629 100 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 17968769 Leverandør VHG Labs Leverandørnr. VHGPTHN500

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Thorium Standard: Th @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Thorium
CAS 7440-29-1
Metoder ICP/ICP-MS
Analyt- eller komponentnavne Thorium
Forsendelsestilstand Room Temperature
Molekylær formel Th
Koncentration 1000 μg/mL
Certificeringer/compliance ISO 17034
Synonym Th
InChI formel InChI=1S/Th
SMIL [Th]
IUPAC navn thorium
Opløsningsmiddel eller Matrix Dilute HNO3
Molekylvægt (g/mol) 232.038
Mængde 500 ml
Formel vægt 232.038 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.