Få mere at vide
Tocris Bioscience™ THIP hydrochloride
Beskrivelse
Systemically active GABAA receptor agonist and GABAA-ρ receptor antagonist. Displays antinociceptive, anticonvulsant and sedative effects. Hypnotic agent that enhances delta activity within non-REM sleep in rats.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | THIP hydrochloride |
| CAS | 85118-33-8 |
| Mængde | 50 mg |
| Molekylær formel | C6H9ClN2O2 |
| Formel vægt | 176.6 |
| Renhed | >98% |
| Molekylvægt (g/mol) | 176.60 |