Få mere at vide
Thiouric Acid Sodium Salt Dihydrate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Thiouric Acid Sodium Salt Dihydrate |
| CAS | 1329805-85-7 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C5 H3 N4 O2 S . Na . 2 H2 O |
| Mængde | 250 mg |
| Synonym | 7,9-Dihydro-6-thioxo-1H-purine-2,8(3H,6H)-dione Sodium Salt Dihydrate, 6-Thio-uric Acid Sodium Dihydrate, 2,8-Dihydroxy-6-mercaptopurine Sodium Dihydrate, 6-Thiouric Acid Sodium Dihydrate, NSC 46380 |
| InChI formel | InChI=1S/C5H4N4O2S.Na.2H2O/c10-4-6-1-2(7-4)8-5(11)9-3(1)12;;;/h(H4,6,7,8,9,10,11,12);;2*1H2/q;+1;;/p-1 |
| SMIL | O.O.[Na+].Oc1nc([S-])c2nc(O)[nH]c2n1 |
| IUPAC navn | sodium;2,8-dihydroxy-9H-purine-6-thiolate;dihydrate |
| Molekylvægt (g/mol) | 242.19 |
| Vis mere |