Få mere at vide
Thiosildenafil, Mikromol™
Artikelnummer. 30458200
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30458200 4 x 25 mg
30049263 25 mg
17970506 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30458200 Leverandør Mikromol Leverandørnr. MM0254.3900254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Thiosildenafil, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Thiosildenafil
CAS 479073-79-5
Analyt- eller komponentnavne Thiosildenafil
Forsendelsestilstand Room Temperature
Molekylær formel C22 H30 N6 O3 S2
Certificeringer/compliance ISO 17025
Synonym 7H-Pyrazolo[4,3-d]pyrimidine-7-thione, 5-[2-ethoxy-5-[(4-methyl-1-piperazinyl)sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-, Piperazine, 1-[[3-(4,7-dihydro-1-methyl-3-propyl-7-thioxo-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-methyl- (9CI), Thiosildenafil
InChI formel InChI=1S/C22H30N6O3S2/c1-5-7-17-19-20(27(4)25-17)22(32)24-21(23-19)16-14-15(8-9-18(16)31-6-2)33(29,30)28-12-10-26(3)11-13-28/h8-9,14H,5-7,10-13H2,1-4H3,(H,23,24,32)
Løsningstype Neat
SMIL CCCc1nn(C)c2C(=S)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(C)CC4
IUPAC navn 5-[2-ethoxy-5-(4-methylpiperazin-1-yl)sulfonylphenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidine-7-thione
Molekylvægt (g/mol) 490.64
Mængde 4 x 25 mg
Formel vægt 490.1821 g/mol