Få mere at vide
Thiorphan Disulfide, TRC
Artikelnummer. 30453778
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30453778 25 mg
30448930 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30453778 Leverandør Toronto Research Chemicals Leverandørnr. TRCT36950525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 123658-06-0
Molekylær formel C24H28N2O6S2
Molekylvægt (g/mol) 504.62
Synonym N,N'-[Dithiobis[1-oxo-2-(phenylmethyl)-3,1-propanediyl]]bis-glycine
SMIL O=C(NCC(O)=O)C(CSSCC(C(NCC(O)=O)=O)CC1=CC=CC=C1)CC2=CC=CC=C2
Tekniske data
Kemisk navn eller materiale Thiorphan Disulfide
Mængde 25 mg
InChI formel InChI=1S/C24H28N2O6S2/c27-21(28)13-25-23(31)19(11-17-7-3-1-4-8-17)15-33-34-16-20(24(32)26-14-22(29)30)12-18-9-5-2-6-10-18/h1-10,19-20H,11-16H2,(H,25,31)(H,26,32)(H,27,28)(H,29,30)