Få mere at vide
Thioridazine-d3 5-Sulfoxide, TRC
Artikelnummer. 30456910
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30456910 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30456910 Leverandør Toronto Research Chemicals Leverandørnr. TRCT3688171MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Thioridazine-d3 5-Sulfoxide
CAS 1330076-56-6
Molekylær formel C21H23D3N2OS2
Mængde 1 mg
Synonym 10-[2-(1-Methyl-2-piperidinyl)ethyl]-2-(methylthio)-10H-phenothiazine-d3 5-Oxide, 2-Methylsulfanyl-10-[2-[1-(trideuteriomethyl)piperidin-2-yl]ethyl]phenothiazine 5-oxide
InChI formel InChI=1S/C21H26N2OS2/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)26(24)21-11-10-17(25-2)15-19(21)23/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3/i1D3
SMIL O=S1C2=CC=CC=C2N(CCC3CCCCN3C([2H])([2H])[2H])C4=C1C=CC(SC)=C4
IUPAC navn 10-(2-(1-(methyl-d3)piperidin-2-yl)ethyl)-2-(methylthio)-10H-phenothiazine 5-oxide
Molekylvægt (g/mol) 389.59
Formel vægt 389.17