missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioredoxin-like 5/TRP14/TXNDC17 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99769IR
Additional Details : Weight : 0.00970kg
Description
Thioredoxin-like 5/TRP14/TXNDC17 Polyclonal specifically detects Thioredoxin-like 5/TRP14/TXNDC17 in Human samples. It is validated for ELISA.Specifications
Thioredoxin-like 5/TRP14/TXNDC17 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
84817 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
DyLight 755 | |
MGC14353, Protein 42-9-9, thioredoxin (Trx)-related protein, 14 kDa, thioredoxin domain containing 17, thioredoxin domain-containing protein 17, thioredoxin-like 5, Thioredoxin-like protein 5, TRP1414 kDa thioredoxin-related protein, TXNL5 | |
Produced in rabbits immunized with purified, recombinant Thioredoxin-like 5/TRP14/TXNDC17 (Q9BRA2; Met1-Asp123). | |
0.1 mL | |
Primary | |
Human | |
Purified |