Få mere at vide
Thioproperazine Dimesilate, Mikromol™
Beskrivelse
Thioproperazine Dimesilate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Thioproperazine Dimesilate |
| CAS | 2347-80-0 |
| Analyt- eller komponentnavne | Thioproperazine Dimesilate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C22 H30 N4 O2 S2 . 2 C H4 O3 S |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 10H-Phenothiazine-2-sulfonamide, N,N-dimethyl-10-[3-(4-methyl-1-piperazinyl)propyl]-, methanesulfonate (1:2), 10H-Phenothiazine-2-sulfonamide, N,N-dimethyl-10-[3-(4-methyl-1-piperazinyl)propyl]-, dimethanesulfonate (9CI), Phenothiazine-2-sulfonamide, N,N-dimethyl-10-[3-(4-methyl-1-piperazinyl)propyl]-, dimethanesulfonate (6CI,7CI,8CI), Mageptyl, Majeptil, Thioperazine dimethanesulfonate, Thioproperazine bismethanesulfonate, Thioproperazine dimesylate, Thioproperazine dimethanesulfonate, Thioproperazine mesylate, Thioproperazine methanesulfonate, Vontil |
| InChI formel | InChI=1S/C22H30N4O2S2.2CH4O3S/c1-23(2)30(27,28)18-9-10-22-20(17-18)26(19-7-4-5-8-21(19)29-22)12-6-11-25-15-13-24(3)14-16-25;2*1-5(2,3)4/h4-5,7-10,17H,6,11-16H2,1-3H3;2*1H3,(H,2,3,4) |
| SMIL | CN(C)S(=O)(=O)c1ccc2Sc3ccccc3N(CCCN4CCN(C)CC4)c2c1.CS(=O)(=O)O.CS(=O)(=O)O |
| IUPAC navn | N,N-dimethyl-10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine-2-sulfonamide;methanesulfonic acid |
| Vis mere |