Få mere at vide
Thiophene-d4, CDN
Artikelnummer. 30322524
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322524 0.5 g 0.5g
30318411 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322524 Leverandør CDN Leverandørnr. CDND28600.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Thiophene-D4
CAS 2036-39-7
Molekylær formel C4 D4 S
Mængde 0.5 g
Synonym Thiophene-2,3,4,5-d4 (ACI), Thiophene-d4 (6 CI, 7 CI, 8 CI, 9 CI), 2,3,4,5-Tetradeuteriothiophene, Perdeuteriothiophene
InChI formel InChI=1 S/C4H4S/c1-2-4-5-3-1/h1-4 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1sc([2 H])c([2 H])c1[2 H]
IUPAC navn 2,3,4,5-tetradeuteriothiophene
Molekylvægt (g/mol) 88.1642
Formel vægt 88.0285 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity