Få mere at vide
Thiophene-2,5-d2, CDN
Artikelnummer. 30312550
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30312550 0.25 g -
30310794 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312550 Leverandør CDN Leverandørnr. CDND28590.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Thiophene-2,5-D2
CAS 2041-42-1
Molekylær formel C4 D2 H2 S
Mængde 0.25 g
Synonym Thiophene-2,5-d2 (6 CI, 7 CI, 8 CI, 9 CI), 2,5-Dideuteriothiophene, Thiophene-2,5-D2
InChI formel InChI=1 S/C4H4S/c1-2-4-5-3-1/h1-4 H/i3D,4 D
SMIL [2 H]c1ccc([2 H])s1
IUPAC navn 2,5-dideuteriothiophene
Molekylvægt (g/mol) 86.152
Formel vægt 86.016 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity