Få mere at vide
Thiophanate-methyl-d6, TRC
Artikelnummer. 30453713
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30453713 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453713 Leverandør Toronto Research Chemicals Leverandørnr. TRCT34481210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Thiophanate-methyl-d6
CAS 1398065-77-4
Molekylær formel C12 H8 D6 N4 O4 S2
Mængde 10 mg
Synonym N,N'-[1,2-Phenylenebis(iminocarbonothioyl)]biscarbamic-d6 Acid C,C'-Dimethyl Ester, 4,4'-o-Phenylenebis[3-thioallophanic-d6 Acid Dimethyl Ester, 1,2-Bis[3-(methoxycarbonyl)-2-thioureido]benzene-d6, 3336G-d6, Caligran-d6, Cercobin M 70-d6, Cercobin Methyl-d6, Cycosin-d6, Dimethyl 4,4'-o-Phenylenebis(3-thioallophanate)-d6, Easout-d6, Enovit M 70-d6, Enovit Super-d6, Enovit-methyl-d6, F 6385-d6, Fungo-d6, Methyl topsin-d6, Methylthiofanate-d6, Methylthiophanate-d6, Metoben-d6, Mildothane-d6, NF 44-d6, NSC 170811-d6, Neotopsin-d6, Thiopan-d6, Thiophanate methyl-d6, Tops-d6, Topsin 500SC-d6, Topsin M 70WP-d6, Topsin MD-d6, Topsin Methyl-d6, o-Bis(3-methoxycarbonyl-2-thioureido)benzene-d6
InChI formel InChI=1S/C12H14N4O4S2/c1-19-11(17)15-9(21)13-7-5-3-4-6-8(7)14-10(22)16-12(18)20-2/h3-6H,1-2H3,(H2,13,15,17,21)(H2,14,16,18,22)/i1D3,2D3
SMIL [2H]C([2H])([2H])OC(=O)NC(=S)Nc1ccccc1NC(=S)NC(=O)OC([2H])([2H])[2H]
IUPAC navn trideuteriomethyl N-[[2-(trideuteriomethoxycarbonylcarbamothioylamino)phenyl]carbamothioyl]carbamate
Molekylvægt (g/mol) 348.43
Formel vægt 348.0833