Få mere at vide
Novus Biologicals™ Thiol Peroxidase Protein
Artikelnummer. 18219128 Shop alle Bio Techne produkter
Mængde:
0.5 mg
100μg
Artikelnummer. 18219128
Brand: Novus Biologicals™ NBP178805100UG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-168 of E.coli Thiol Peroxidase The Recombinant E. coli Thiol Peroxidase Protein is derived from E. coli. The Recombinant E. coli Thiol Peroxidase Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Tekniske data
Adgangsnummer NP_415840
Koncentration 1mg/mL
Til brug med (applikation) SDS-PAGE
Formulering 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 2mM DTT, 0.1M NaCl.
Molekylvægt (g/mol) 19.9kDa
Oprensningsmetode SDS-PAGE
Mængde 100μg
Kilde Recombinant E. coli TPX protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen TPX, 1-168aa, E. coli, His tag, E. coli (NP_415840).
Opbevaringskrav Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatorisk status RUO
Fælles navn Thiol Peroxidase
Konjugeret Unconjugated
Rekombinant Recombinant
Renhed eller kvalitetsklasse >95%