missing translation for 'onlineSavingsMsg'
Få mere at vide

Thiodigalactoside, TRC

Artikelnummer. 30452282
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30452282 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452282 Supplier Toronto Research Chemicals Supplier No. TRCT34459525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Thiodigalactoside
CAS 51555-87-4
Molekylær formel C12H22O10S
Mængde 25 mg
Synonym Galactopyranoside','β-D-galactopyranosyl 1-thio-','β-D- (8CI), β-D-Galactopyranosyl 1-thio-β-D-galactopyranoside (ACI), D-Galactopyranosylthio-β-D-galactopyranoside, Galactosyl 1-thiogalactoside, Thio di-β-D-galactopyranoside, Thiodigalactoside, β-D-Galactopyranosyl-β′-D-galactopyranosyl-sulfide, β-D-Galactosyl 1-thio-β-D-galactoside
InChI formel InChI=1S/C12H22O10S/c13-1-3-5(15)7(17)9(19)11(21-3)23-12-10(20)8(18)6(16)4(2-14)22-12/h3-20H,1-2H2/t3-,4-,5+,6+,7+,8+,9-,10-,11+,12+/m1/s1
SMIL OC[C@H]([C@@H]([C@@H]([C@H]1O)O)O)O[C@H]1S[C@@H]2O[C@@H]([C@@H]([C@@H]([C@H]2O)O)O)CO
IUPAC navn (2R,2'R,3R,3'R,4S,4'S,5R,5'R,6S,6'S)-6,6'-thiobis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol)
Molekylvægt (g/mol) 358.36
Formel vægt 358.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.