Få mere at vide
Thiodigalactoside, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Thiodigalactoside |
| CAS | 51555-87-4 |
| Molekylær formel | C12H22O10S |
| Mængde | 100 mg |
| Synonym | Galactopyranoside','β-D-galactopyranosyl 1-thio-','β-D- (8CI), β-D-Galactopyranosyl 1-thio-β-D-galactopyranoside (ACI), D-Galactopyranosylthio-β-D-galactopyranoside, Galactosyl 1-thiogalactoside, Thio di-β-D-galactopyranoside, Thiodigalactoside, β-D-Galactopyranosyl-β′-D-galactopyranosyl-sulfide, β-D-Galactosyl 1-thio-β-D-galactoside |
| InChI formel | InChI=1S/C12H22O10S/c13-1-3-5(15)7(17)9(19)11(21-3)23-12-10(20)8(18)6(16)4(2-14)22-12/h3-20H,1-2H2/t3-,4-,5+,6+,7+,8+,9-,10-,11+,12+/m1/s1 |
| SMIL | OC[C@H]([C@@H]([C@@H]([C@H]1O)O)O)O[C@H]1S[C@@H]2O[C@@H]([C@@H]([C@@H]([C@H]2O)O)O)CO |
| IUPAC navn | (2R,2'R,3R,3'R,4S,4'S,5R,5'R,6S,6'S)-6,6'-thiobis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) |
| Molekylvægt (g/mol) | 358.36 |
| Formel vægt | 358.06 |