missing translation for 'onlineSavingsMsg'
Få mere at vide

Thiocolchicoside Tetraacetate, TRC

Product Code. 30451971
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30451971 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451971 Supplier Toronto Research Chemicals Supplier No. TRCT344585100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Thiocolchicoside Tetraacetate
CAS 163956-01-2
Molekylær formel C35H41NO14S
Mængde 100 mg
Synonym N-[(7S)-5,6,7,9-Tetrahydro-1,2-dimethoxy-10-(methylthio)-9-oxo-3-[(2,3,4,6-tetra- O-acetyl-beta-D-glucopyranosyl)oxy]benzo[a]heptalen-7-yl]acetamide, (S)-N-[5,6,7,9-tetrahydro-1,2-dimethoxy-10-(methylthio)-9-oxo-3-[(2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosyl)oxy]benzo[a]heptalen-7-yl]acetamide, Thiocolchicoside Tetraacetate, Tetra-O-acetylthiocolchicoside
InChI formel InChI=1S/C35H41NO14S/c1-16(37)36-24-11-9-21-13-26(30(43-6)32(44-7)29(21)22-10-12-28(51-8)25(42)14-23(22)24)49-35-34(48-20(5)41)33(47-19(4)40)31(46-18(3)39)27(50-35)15-45-17(2)38/h10,12-14,24,27,31,33-35H,9,11,15H2,1-8H3,(H,36,37)/t24-,27+,31+,33-,34+,35+/m0/s1
SMIL COc1c(O[C@@H]2O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]2OC(=O)C)cc3CC[C@H](NC(=O)C)C4=CC(=O)C(=CC=C4c3c1OC)SC
IUPAC navn [(2R,3R,4S,5R,6S)-6-[[(7S)-7-acetamido-1,2-dimethoxy-10-methylsulfanyl-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-3-yl]oxy]-3,4,5-triacetyloxyoxan-2-yl]methyl acetate
Molekylvægt (g/mol) 731.76
Formel vægt 731.2248
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.