Få mere at vide
Thiobenzamide-2,3,4,5,6-d5, CDN
Artikelnummer. 30321621
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321621 0.5 g
30313921 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321621 Leverandør CDN Leverandørnr. CDND60900.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Thiobenzamide-2,3,4,5,6-D5
CAS 1219804-59-7
Molekylær formel C7 D5 H2 N S
Mængde 0.5 g
Synonym Benzene-2,3,4,5,6-d5-carbothioamide (ACI), Aminothiocarbonylbenzene-2,3,4,5,6-d5, Benzene-d5-thiocarboxamide, Benzothioamide-d5, Thiobenzamide-d5, Thiobenzamide-2,3,4,5,6-D5
InChI formel InChI=1 S/C7H7NS/c8-7(9)6-4-2-1-3-5-6/h1-5 H,(H2,8,9)/i1D,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=S)N)c([2 H])c1[2 H]
IUPAC navn 2,3,4,5,6-pentadeuteriobenzenecarbothioamide
Molekylvægt (g/mol) 142.233
Formel vægt 142.061 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity