Få mere at vide
Thiobencarb, TRC
Artikelnummer. 30453788
Skift visning
Mængde:
1 g
2.5 g
10 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30453788 1 g -
30456004 10 g -
30453186 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30453788 Leverandør Toronto Research Chemicals Leverandørnr. TRCT3442551G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 28249-77-6
Molekylær formel C12 H16 Cl N O S
Molekylvægt (g/mol) 257.78
Synonym Carbamothioic acid, N,N-diethyl-, S-[(4-chlorophenyl)methyl] ester (ACI), Carbamic acid, diethylthio-, S-(p-chlorobenzyl) ester (8CI), Carbamothioic acid, diethyl-, S-[(4-chlorophenyl)methyl] ester (9CI), α-Toluenethiol, p-chloro-, diethylcarbamate (8CI), B 3015, B 3015 (pesticide), Bencarb, Benthiocarb, Bolero, Bolero 8EC, IMC 3950, p-Chlorobenzyl diethylthiolcarbamate, p-Chlorobenzyl N,N-diethylthiolcarbamate, S-(4-Chlorobenzyl) diethylthiocarbamate, S-(4-Chlorobenzyl) diethylthiolcarbamate, S-(4-Chlorobenzyl) N,N-diethylthiocarbamate, S-(p-Chlorobenzyl) N,N-diethylthiocarbamate, S-(p-Chlorobenzyl) N,N-diethylthiolocarbamate, S-4-Chlorobenzyl N,N-diethylthiolcarbamate, S-p-Chlorobenzyl diethylthiocarbamate, Sadan, Saturn, Saturn (pesticide), Thiobencarb
IUPAC navn S-[(4-chlorophenyl)methyl] N,N-diethylcarbamothioate
SMIL CCN(CC)C(=O)SCc1ccc(Cl)cc1
Tekniske data
Kemisk navn eller materiale Thiobencarb
Mængde 1 g
InChI formel InChI=1S/C12H16ClNOS/c1-3-14(4-2)12(15)16-9-10-5-7-11(13)8-6-10/h5-8H,3-4,9H2,1-2H3
Formel vægt 257.0641