Få mere at vide
Thiobencarb-d10 (diethyl-d10), TRC
Artikelnummer. 30454577
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30454577 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454577 Leverandør Toronto Research Chemicals Leverandørnr. TRCT3442565MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Thiobencarb diethyl-d10
CAS 1219804-12-2
Molekylær formel C12 D10 H6 Cl N O S
Mængde 5 mg
Synonym Carbamothioic acid','N,N-di(ethyl-1,1,2,2,2-d5)-','S-[(4-chlorophenyl)methyl] ester (ACI), S-[(4-Chlorophenyl)methyl] N,N-di(ethyl-1,1,2,2,2-d5)carbamothioate (ACI), Thiobencarb-d10, N,N-Di(ethyl-1,1,2,2,2-d5)-S-[(4-Chlorophenyl)methyl] Ester Carbamothioic Acid
InChI formel InChI=1S/C12H16ClNOS/c1-3-14(4-2)12(15)16-9-10-5-7-11(13)8-6-10/h5-8H,3-4,9H2,1-2H3/i1D3,2D3,3D2,4D2
SMIL [2H]C([2H])([2H])C([2H])([2H])N(C(=O)SCc1ccc(Cl)cc1)C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn S-[(4-chlorophenyl)methyl] N,N-bis(1,1,2,2,2-pentadeuterioethyl)carbamothioate
Molekylvægt (g/mol) 267.84
Formel vægt 267.1269