Få mere at vide
Thiobencarb-d10 (diethyl-d10), CDN
Artikelnummer. 30328627
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328627 0.01 g
30315644 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328627 Leverandør CDN Leverandørnr. CDND64950.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Thiobencarb diethyl-d10
CAS 1219804-12-2
Molekylær formel C12 D10 H6 Cl N O S
Mængde 0.01 g
Synonym N,N-Di(ethyl-1,1,2,2,2-d5)-S-[(4-Chlorophenyl)methyl] Ester Carbamothioic Acid
InChI formel InChI=1 S/C12H16ClNOS/c1-3-14(4-2)12(15)16-9-10-5-7-11(13)8-6-10/h5-8 H,3-4,9H2,1-2H3/i1D3,2D3,3D2,4D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])N(C(=O)SCc1ccc(Cl)cc1)C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn S-[(4-chlorophenyl)methyl] N,N-bis(1,1,2,2,2-pentadeuterioethyl)carbamothioate
Molekylvægt (g/mol) 267.84
Formel vægt 267.1269 g/mol
Procent renhed 99 atom % D, min 97% Chemical Purity