Få mere at vide
Fisherbrand™ Thin-Walled PCR Tubes
Shop alle Fisherbrand produkter
Farve:
Diverse
Neutral
Lukketype:
Flad kasket
Kuppelformet kasket
Volumen (metrisk):
0,2 ml
0,5 ml
Format:
8-strips
Individuelt
Pakningsstørrelse:
Pakke med 1000
Pakke med 960
Beskrivelse
- Available in mixed or natural colors
- Available in individual tubes or strips
- Available with a variety of caps - domed, flat, strip
Tekniske data
Tekniske data
| Farve | Diverse |
| Volumen (metrisk) | 0,2 ml |
| Lukningsmateriale | Polypropylene |
| Til brug med (udstyr) | PCR |
| Lukketype | Kuppelformet kasket |
| Format | 8-strips |
| Produkttype | PCR Tube |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.