missing translation for 'onlineSavingsMsg'
Learn More

Thieno[3,2-c]pyridine, TRC

Product Code. 30455513
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30455513 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455513 Supplier Toronto Research Chemicals Supplier No. TRCT344320100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Thieno[3,2-c]pyridine
CAS 272-14-0
Molekylær formel C7 H5 N S
Mængde 100 mg
Synonym Thieno[3,2-c]pyridine, Ticlopidine Hydrochloride Imp. A (EP)
InChI formel InChI=1S/C7H5NS/c1-3-8-5-6-2-4-9-7(1)6/h1-5H
SMIL c1cc2sccc2cn1
IUPAC navn thieno[3,2-c]pyridine
Molekylvægt (g/mol) 135.19
Formel vægt 135.0143
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.