missing translation for 'onlineSavingsMsg'
Learn More

Thiazole-2-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-7055120
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
14190-59-1
C4H3NO2S
129.133
MFCD02094169
IJVLVRYLIMQVDD-UHFFFAOYSA-N
thiazole-2-carboxylic acid, 2-thiazolecarboxylic acid, thiazole-2-carboxylicacid, 2-carboxy-1,3-thiazole, thiazolecarboxylic acid, pubchem16257, thiazole carboxylic acid, 2-carboxythiazole, acmc-209cn5
2762733
1,3-thiazole-2-carboxylic acid
C1=CSC(=N1)C(=O)O
This item is not returnable. View return policy
14190-59-1
C4H3NO2S
129.133
MFCD02094169
IJVLVRYLIMQVDD-UHFFFAOYSA-N
thiazole-2-carboxylic acid, 2-thiazolecarboxylic acid, thiazole-2-carboxylicacid, 2-carboxy-1,3-thiazole, thiazolecarboxylic acid, pubchem16257, thiazole carboxylic acid, 2-carboxythiazole, acmc-209cn5
2762733
1,3-thiazole-2-carboxylic acid
C1=CSC(=N1)C(=O)O

Thiazole-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazole-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 129.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Thiazole-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'