missing translation for 'onlineSavingsMsg'
Learn More

Thianaphthene, 97%, Thermo Scientific Chemicals

Catalog Number p-3737541
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
95-15-8
C8H6S
134.2
MFCD00005864
FCEHBMOGCRZNNI-UHFFFAOYSA-N
benzo b thiophene, thianaphthene, benzothiophene, benzothiofuran, thionaphthene, 1-thiaindene, thianaphthen, benzothiophen, thianaphtene, 1-benzothiophen
7221
CHEBI:35858
1-benzothiophene
C1=CC=C2C(=C1)C=CS2
This item is not returnable. View return policy
95-15-8
C8H6S
134.2
MFCD00005864
FCEHBMOGCRZNNI-UHFFFAOYSA-N
benzo b thiophene, thianaphthene, benzothiophene, benzothiofuran, thionaphthene, 1-thiaindene, thianaphthen, benzothiophen, thianaphtene, 1-benzothiophen
7221
CHEBI:35858
1-benzothiophene
C1=CC=C2C(=C1)C=CS2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 28.0°C to 32.0°C
Color Pink to White or Yellow
Density 1.1400g/mL
Boiling Point 221.0°C to 222.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 17, 59
Merck Index 15, 9455
Specific Gravity 1.14
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in usual organic solvents, soluble in ethanol, ethylether, acetone
Formula Weight 134.2
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material Thianaphthene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Thianaphthene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'