Få mere at vide
Thiamine Triphosphate, TRC
Artikelnummer. 30137928
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30137928 25 mg -
30009301 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137928 Leverandør Toronto Research Chemicals Leverandørnr. TRCT34410025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3475-65-8
Molekylær formel C12H19N4O10P3S
Molekylvægt (g/mol) 504.29
Synonym Thiamine hydroxide, tetrahydrogen triphosphate (ester), inner salt (8CI), Thiamine, triphosphate (7CI), Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-(4,6,8,8-tetrahydroxy-3,5,7-trioxa-4,6,8-triphosphaoct-1-yl)-, inner salt, P,P′,P′′-trioxide (ZCI), Thiamine triphosphoric acid ester
IUPAC navn 3-((4-amino-2-methylpyrimidin-5-yl)methyl)-5-(2-((hydroxy((hydroxy((hydroxyoxidophosphoryl)oxy)phosphoryl)oxy)phosphoryl)oxy)ethyl)-4-methylthiazol-3-ium
SMIL [O-]P(O)(OP(O)(OP(O)(OCCC1=C([N+](CC2=CN=C(N=C2N)C)=CS1)C)=O)=O)=O
Tekniske data
Kemisk navn eller materiale Thiamine Triphosphate
Mængde 25 mg
InChI formel InChI=1S/C12H19N4O10P3S/c1-8-11(30-7-16(8)6-10-5-14-9(2)15-12(10)13)3-4-24-28(20,21)26-29(22,23)25-27(17,18)19/h5,7H,3-4,6H2,1-2H3,(H5-,13,14,15,17,18,19,20,21,22,23)
Formel vægt 504