Få mere at vide
Thiamethoxam-d4, TRC
Artikelnummer. 30454911
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30454911 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454911 Leverandør Toronto Research Chemicals Leverandørnr. TRCT34418225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale ThiamethoxamD4(oxadiazineD4)
CAS 1331642-98-8
Renhedsgrad noter HPLC
Molekylær formel C8 2H4 H6 Cl N5 O3 S
Mængde 25 mg
Synonym 4H-1,3,5-Oxadiazin-(2,2,6,6-D4)-4-imine','3-[(2-chloro-5-thiazolyl)methyl]tetrahydro-5-methyl-N-nitro-
InChI formel InChI=1S/C8H10ClN5O3S/c1-12-4-17-5-13(8(12)11-14(15)16)3-6-2-10-7(9)18-6/h2H,3-5H2,1H3/i4D2,5D2
SMIL [2H]C1([2H])OC([2H])([2H])N(Cc2cnc(Cl)s2)C(=N[N+](=O)[O-])N1C
IUPAC navn N-[3-[(2-chloro-1,3-thiazol-5-yl)methyl]-2,2,6,6-tetradeuterio-5-methyl-1,3,5-oxadiazinan-4-ylidene]nitramide
Molekylvægt (g/mol) 295.74
Formel vægt 295.0444
Procent renhed >95