Få mere at vide
Thiamazole EP Impurity C (as Hydrochloride), Mikromol™
Beskrivelse
Thiamazole EP Impurity C (as Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1-Methyl-2-(methylsulphanyl)-1H-imidazole Hydrochloride |
| CAS | 858220-92-5 |
| Analyt- eller komponentnavne | 1-Methyl-2-(methylsulphanyl)-1H-imidazole Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C5 H8 N2 S . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Thiamazole Imp. C (EP), Methimazole USP Related Compound C, Methimazole USP RC C, 1-Methyl-2-(methylsulphanyl)-1H-imidazole Hydrochloride, Thiamazole Imp. C (EP) as Hydrochloride, Methimazole Related Compound C as Hydrochloride, Thiamazole Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C5H8N2S.ClH/c1-7-4-3-6-5(7)8-2;/h3-4H,1-2H3;1H |
| SMIL | Cl.CSc1nccn1C |
| IUPAC navn | 1-methyl-2-methylsulfanylimidazole;hydrochloride |
| Vis mere |