Få mere at vide
Thiabendazole-d4 (Major), TRC
Artikelnummer. 30454149
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30454149 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454149 Leverandør Toronto Research Chemicals Leverandørnr. TRCT3441521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Thiabendazole-d4
CAS 1190007-20-5
Renhedsgrad noter HPLC
Molekylær formel C10 D4 H3 N3 S
Mængde 1 mg
Synonym 4,5,6,7-Tetradeuterio-2-(1,3-thiazol-4-yl)-1H-benzimidazole, 2-(1,3-Thiazol-4-yl)-1H-benzimidazole-4,5,6,7-d4, Thiabendazole-d4, Tiabendazole-d4, Tiabendazole-D4 (benzimidazole-4,5,6,7-D4)','Thiabendazole D4 (benzimidazol 4,5,6,7 D4)
InChI formel InChI=1S/C10H7N3S/c1-2-4-8-7(3-1)12-10(13-8)9-5-14-6-11-9/h1-6H,(H,12,13)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c2[nH]c(nc2c1[2H])c3cscn3
IUPAC navn 4-(4,5,6,7-tetradeuterio-1H-benzimidazol-2-yl)-1,3-thiazole
Molekylvægt (g/mol) 205.272
Formel vægt 205.061
Procent renhed >95