Få mere at vide
Thiabendazole-d4 (benzimidazole-4,5,6,7-d4), CDN
Artikelnummer. 30326850
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326850 0.005 g
30320897 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326850 Leverandør CDN Leverandørnr. CDND73150.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Thiabendazole-d4
CAS 1190007-20-5
Molekylær formel C10 D4 H3 N3 S
Mængde 0.005 g
Synonym 4,5,6,7-Tetradeuterio-2-(1,3-thiazol-4-yl)-1 H-benzimidazole, 2-(1,3-Thiazol-4-yl)-1 H-benzimidazole-4,5,6,7-d4, Thiabendazole-d4, Tiabendazole-d4, Tiabendazole-D4 (benzimidazole-4,5,6,7-D4), Thiabendazole D4 (benzimidazol 4,5,6,7 D4)
InChI formel InChI=1 S/C10H7N3S/c1-2-4-8-7(3-1)12-10(13-8)9-5-14-6-11-9/h1-6 H,(H,12,13)/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c([2 H])c2[nH]c(nc2c1[2 H])c3cscn3
IUPAC navn 4-(4,5,6,7-tetradeuterio-1 H-benzimidazol-2-yl)-1,3-thiazole
Molekylvægt (g/mol) 205.272
Formel vægt 205.061 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity