Learn More
VEE GEE Thermometer, Encapsulated in FEP
The encapsulated glass thermometer is designed to safeguard against broken glass and liquid. It contains a biodegradable Safety Blue fill, offering mercury-like properties minus the associated risks of contamination and hazards.
Brand:  VEE GEE 80903E
 View more versions of this product
Product Code. 17811214
  • 530.00 DKK / Each
Description
  • FEP encapsulation- in the event of breakage the FEP coating contains the broken glass and liquid, preventing injury or contamination
  • Feature Safety Blue fill against a white background for easy reading
  • Accuracy is equal to one subdivision
  • Immersion: Total or Partial
Specifications
Specifications
Safety Blue
Partial Immersion
Glass
Safety Blue
Serialized Glass Thermometer
148.9°C
-17.8°C
-17.8 to 148.9°C
1.1°C (2°F)
1.5 cm
Laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications
30.48 cm
Immersion/General Purpose (Tapered)
5 g
300°F
0°F
0 to 300°F
±1 subdivision
To immersion line
Certificates
Promotions
Promotions
Product Content Correction
Product Title
VEE GEE Thermometer, Encapsulated in FEP > Immersion Length: To immersion line, Immersion Type: Partial Immersion, Temperature Range: 0 to 300°F