Learn More
Unity Lab Services™ Thermo Scientific Smart-Vue Pro Remote Monitoring System Qualification Service
Product Code. 16628035
Change view
Click to view available options
For Use With (Equipment):
Smart-Vue Pro Remote Monitoring System
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
16628035 Smart-Vue Pro Remote Monitoring System Each
1 options
Product Code. 16628035 Supplier Unity Lab Services™ Supplier No. IOQPCKE89003675
 Restricted Item
Forventet forsendelse: 13-01-2027
Stay compliant with our industry exclusive no charge requalification guarantee included with each Thermo Scientific validation service for Smart-Vue Pro Remote Monitoring System
IQ
  • Unpacking, Assembly and Installation Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
  • Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ
  • System Component Information Detailed configuration information for each system component have been recorded.
  • OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
  • Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment.
Key Tests: Handling Alarms, Alarm Management, Voice Call, Alerts, Managing Readings, User & Call Group Management, Reports/Printouts
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Smart-Vue Pro Remote Monitoring System
Product Type IQ/OQ Service
Location Contact your local sales representative for a list of coverage areas.