Learn More
Thermo Scientific™ Nunc™ Open Well Upper Structures
Versatile units in a choice of two materials
Brand: Thermo Scientific™ 256665
4954.08 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Versatile units in a choice of two materials
Used for bonding to a specialty base material, holding PCR plates, adding agar to each well, or other purposes
- Nonsterile, black
- 96- well formats available
- Polystyrene
- Nonsterile
- Without lid
Specifications
Black | |
96 | |
100 | |
Case of 100 |
Polystyrene | |
Non-sterile | |
96-Well Plate |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1286021 | Certificate of Compliance | 11/12/2024 | 256665 |
1418367 | Certificate of Compliance | 06/12/2024 | 256665 |
1401115 | Certificate of Compliance | 08/03/2024 | 256665 |
1315712 | Certificate of Compliance | 29/04/2021 | 256665 |
1292016 | Certificate of Compliance | 25/08/2020 | 256665 |