Få mere at vide
Thermo Scientific™ HyPURITY™ C8 Guard Cartridges
Artikelnummer. 10714437 Shop alle Thermo Scientific produkter
:
Particle Size: 5μm; 10L x 2.1mm I.D.
Artikelnummer. 10714437
Brand: Thermo Scientific™ 22205012101
Kræver
Thermo Scientific UNIGUARD direct connection or stand alone holder, sold separately.
Drop-in guard cartridges offer convenience, economy and effective protection for extending column lifetime
  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • High quality, highly pure silica columns
TRUSTED_SUSTAINABILITY
Tekniske data
Partikelstørrelse 5 μm
Længde (metrisk) 10 mm
Diameter (metrisk) Indvendig 2.1 mm
Stationær fase C8