Få mere at vide
Thermo Scientific™ HyPURITY™ C4 Guard Cartridge
Shop alle Thermo Scientific produkter
:
Particle Size: 5μm; 10L x 3.0mm I.D.
Beskrivelse
Kræver
Thermo Scientific UNIGUARD direct connection or stand alone holder, sold separately.
- The 10mm design offers maximum protection with minimal increase in retention
- Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
- High quality, highly pure silica columns
Tekniske data
Tekniske data
| Partikelstørrelse | 5 μm |
| Længde (metrisk) | 10 mm |
| Diameter (metrisk) Indvendig | 3 mm |
| Stationær fase | C4 |