Få mere at vide
Thermo Scientific™ Hypersil GOLD™ C8 Javelin™ Guard Columns
Artikelnummer. 11538690 Shop alle Thermo Scientific produkter
Diameter (metrisk) Indvendig:
3 mm
Partikelstørrelse:
5 μm
Pakningsstørrelse:
Pakke med 4
Artikelnummer. 11538690
Brand: Thermo Scientific™ 25205013006
Javelin guard columns offer convenient, easy to use protection for analytical HPLC columns
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of Hypersil GOLD C8 Solid Phase
  • Hypersil GOLD C8 columns provide similar selectivity to C18 columns but with reduced retention
  • Lower hydrophobicity
  • Faster separations
  • Excellent peak shape
  • High efficiency
  • Outstanding sensitivity
Ordreinfo
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.
TRUSTED_SUSTAINABILITY
Tekniske data
Partikelstørrelse 5 μm
Længde (metrisk) 10 mm
Diameter (metrisk) Indvendig 3 mm
Stationær fase C8