Learn More
Thermo Scientific™ HPLC Syringes for CTC Instruments
HPLC syringes offer easy, accurate, and reproducible injections.
Brand:  Thermo Scientific™ 365HL331
 View more versions of this product
Product Code. 13496866
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
22 G
Gastight HPLC Syringe
100 μL
Fixed
1.9 in.
Ea.
CTC Instruments
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ HPLC Syringes for CTC Instruments > Gas tight; 100μL; 51mm; 22 gauge; Fixed needle