Få mere at vide
Thermo Scientific™ Buffers for MagJET™ Blood gDNA Kit
Artikelnummer. 13515200 Shop alle Thermo Scientific produkter
Mængde:
40 mL
Pakningsstørrelse:
40ml
Artikelnummer. 13515200
Brand: Thermo Scientific™ R2371
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Wash lysed samples to remove any contaminants or inhibitors from the isolated genomic DNA. Thermo Scientific™ MagJET™ Wash Buffer 2 (concentrated) is a component of the MagJET™ Whole Blood gDNA Kits (K2741/2742) and may be purchased separately in 40mL aliquots.
The Thermo Scientific™ MagJET™ Whole Blood Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from fresh or frozen whole blood treated with EDTA or citrate, as well as blood fractions. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
TRUSTED_SUSTAINABILITY
Tekniske data
Til brug med (udstyr) MagJET Plant gDNA Kit
Mængde 40 mL