Få mere at vide
Thermo Scientific™ BioBasic™ SEC 60A HPLC Columns
Shop alle Thermo Scientific produkter
Diameter (metrisk):
2.1 mm
4.6 mm
Pakningsstørrelse:
Hver
Beskrivelse
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straghtforward method development, simple mobile phases
- 60Å pore size
- Particle size: 5μm
Ordreinfo
For information on bulk quantities, please inquire.
Tekniske data
Tekniske data
| Kolonneformat | Analytical Column |
| Diameter (metrisk) | 4.6 mm |
| Længde (metrisk) | 150 mm |
| Partikelstørrelse | 5 μm |
| Porestørrelse | 60 Å |
| Stationær fase | Proprietary |
| Produktlinje | BioBasic |
| Emballagemateriale | Silica |
| Kolonnetype | Mixed Mode |
| Fase | Size Exclusion and Gel Permeation |