Learn More
Thermo Scientific™ BioBasic™ SEC 300A HPLC Columns
Outstanding size separation of water soluble compounds
5267.28 DKK
Specifications
Length (Metric) | 250 mm |
---|---|
Particle Size | 5 μm |
Pore Size | 300 Å |
Stationary Phase | Proprietary |
Packing Material | Silica |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Stationary Phase | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Stationary Phase | Price | Quantity & Availability | |||
11582811
|
Thermo Scientific™
73505-254630 |
4.6 mm | 250 mm | Proprietary |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straghtforward method development, simple mobile phases
- 300Å pore size
Order Info
For information on bulk quantities, please inquire.
Specifications
250 mm | |
300 Å | |
Silica |
5 μm | |
Proprietary |