Learn More
Thermo Scientific™ ART™ Solvent Safe Carbon Filtered Pipette Tips
For acids, bases and aggressive organic solvents
Brand: Thermo Scientific™ 5079EB
580.02 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.56700kg
Description
For acids, bases and aggressive organic solvents
Revolutionary filter material handles the pipetting rigors of Combinatorial Chemistry. Unique use of the ART™ filter with Folded Activated Carbon offers both pipetter and sample protection against destructive carryover aerosols and vapors from volatile organic solvents, all without compromising pipetting accuracy and precision.
- For manual pipetting of organic solvents
- Utilizes new formula of the ART (aerosol resistant tip) filter tip
- Helps to eliminate pipetting failures and critical inaccuracies caused by strong acids, bases and aggressive organic solvents
- Proprietary blend of Folded Activated Carbon with a 40-micron polyethylene filter provides effective solvent protection
Specifications
100 to 1000 μL | |
Non-sterile | |
Universal Fit | |
ART |
Filtered | |
Bulk | |
Universal Fit | |
Carbon with Polyethylene Filter |