Få mere at vide
Thermo Scientific™ SuperPlate PCR Plates, 96-well
Artikelnummer. 10124183
Farve:
Blå
Gul
Hvid
Klar
Stregkode:
Ingen stregkode
Inkluderer stregkode
Pakningsstørrelse:
Pakke med 25
Nederdel stil:
Fuld
Semi-
Produkttype:
Hævet dæk
Artikelnummer. 10124183
Brand: Thermo Scientific™ AB2100
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
96-well PCR plates with a flat deck for improved sealing in PCR and qPCR applications.
The Thermo Scientific 96-Well Semi-Skirted Flat Deck PCR Plates are compatible with most thermal cyclers and ABI platforms, including sequencers, with no adapters necessary. The Thermo Scientific PCR plastics are designed, manufactured, and tested to ensure PCR performance. The plates have 0.3 mL maximum well volume when used with adhesive and heat seals.
The flat plate deck design ensures easy handling, and the plates feature a raised rim to prevent evaporation. The 96-Well Semi-Skirted Flat Deck PCR Plates have a cut corner at the A12 location and have thin well walls for consistent heat transfer.
Highlights
• Directly compatible with all major major thermal cyclers, including ABI platforms and sequencers with no adapters necessary
• Flat plate deck facilitates sealing and handling
• Available as SuperPlate providing 4x more rigidity for superior robotic handling
• Raised rim design around each well enables secure sealing and safeguards against evaporation
• Uniform, thin well walls deliver maximum and consistent heat transfer
• Black lettered versions feature black alpha-numeric lettering to allow for easy well identification
• White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
• Cut corner: A12
Applications
• PCR
• qPCR/Real-Time PCR
• Sequencing
Hazardous:No
>Storage Condition:Ambient
TRUSTED_SUSTAINABILITY
Tekniske data
Stregkode Ingen stregkode
Certificeringer/compliance DNA, DNase, RNase-free
Format Standard
Array 8 x 12
Farve Klar
Materiale Polypropylene
Antal Wells 96
Til brug med (applikation) For improved sealing in PCR and qPCR applications
Kantet Yes
Vel Farve Clear