Få mere at vide
Brady™ Thermal Transfer Printable Labels, Paper
Beskrivelse
- Ideal label sizing for logistical applications.
Tekniske data
Tekniske data
| Klæbende type | Permanent Acrylic |
| Belægning | None |
| Farve | White |
| Slutte | Matte |
| Format | Die-cut |
| Materiale | Paper |
| Printbare sider | Single-side Printable |
| Print teknologi | Thermal Transfer |
| Produkttype | Label |
| Sterilitet | Non-sterile |
| Vis mere |