missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Therma Welder Gloves

Catalog Number 12637523
Click to view available options
Size: 11
This item is not returnable. View return policy
You may only order up to the available stock. For more details call customer services on +45 70 27 99 20.
This item is not returnable. View return policy
You may only order up to the available stock. For more details call customer services on +45 70 27 99 20.

Especially suitable for TIG welding operations, these water-repellent gloves are constructed of heat-resistant grain leather. Honeywell Safety Products™ Therma Welder Gloves are also stitched with Kevlar™ thread for additional durability and heat resistance.

  • Suitable for TIG welding with high dexterity.
  • Heat-resistant grain leather construction.
  • Water-repellent treatment for use in wet environments.
  • Split leather cuff to protect the forearm.
  • U.S. assembly.
  • Gunn cut, winged thumb with yoke, crispin type.
  • Stitched with sturdy and heat-resistant Kevlar™ thread.
TRUSTED_SUSTAINABILITY
Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EN 407-04 Thermical hazards, EN 12477-02 Welding Type B, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number 0075/797/162/02/07/0054, EC Attestation Number 0075/797/162/02/07/0054
Classification Dexterity: 5
Cuff Material Split Leather
Cut Resistance Rating 1
Material Grain Leather, Cowhide Leather, Aramid or Dupont™ Kevlar™ aramid fibers
Product Type Crispin Welding Gloves (Type B)
Size 11
Thickness (Metric) 0.8/1.1 mm (Back)
For Use With (Application) Welding
Puncture Rating 1
Quantity 10Pair/PK
Resistant To Heat
Tear Rating 2
Length (Metric) 39 cm
Length (Metric) Cuff 20 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'