Få mere at vide
Theophylline-13C2d6, TRC
Artikelnummer. 30452272
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30452272 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452272 Leverandør Toronto Research Chemicals Leverandørnr. TRCT34385410MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Theophylline-13C2d6
CAS 1782458-84-7
Molekylær formel C513C2H2D6N4O2
Mængde 10 mg
Synonym 3,9-Dihydro-1,3-dimethyl-1H-purine-2,6-dione-13C2d6, 3,7-Dihydro-1,3-dimethyl- 1H-purine-2,6-dione-13C2d6, 1,3-Dimethylxanthine-13C2d6, Accurbron-13C2d6, Aerobin-13C2d6, Armophylline-13C2d6, Asmax-13C2d6, Bronkodyl-13C2d6, Theophyl-13C2d6, Theoplus-13C2d6, Theostat-13C2d6, Unifyl-13C2d6, Unilong-13C2d6, Xanthium-13C2d6
InChI formel InChI=1S/C7H8N4O2/c1-10-5-4(8-3-9-5)6(12)11(2)7(10)13/h3H,1-2H3,(H,8,9)/i1+1D3,2+1D3
SMIL O=C1C2=C(N=CN2)N([13C]([2H])([2H])[2H])C(N1[13C]([2H])([2H])[2H])=O
Molekylvægt (g/mol) 188.19
Formel vægt 188.1091