Få mere at vide
Theobromine-d6, TRC
Artikelnummer. 30452197
Skift visning
Mængde:
2 mg
Artikelnummer. Mængde
30452197 2 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452197 Leverandør Toronto Research Chemicals Leverandørnr. TRCT3438022MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Theobromine-d6
CAS 117490-40-1
Renhedsgrad noter HPLC
Molekylær formel C7 2H6 H2 N4 O2
Mængde 2 mg
Synonym Theophylline-Ethylenediamine EP Impurity G-d6 (Caffeine EP Impurity D-d6','Theobromine-d6)
InChI formel InChI=1S/C7H8N4O2/c1-10-3-8-5-4(10)6(12)9-7(13)11(5)2/h3H,1-2H3,(H,9,12,13)/i1D3,2D3
SMIL [2H]C([2H])([2H])N1C(=O)NC(=O)c2c1ncn2C([2H])([2H])[2H]
IUPAC navn 3,7-bis(trideuteriomethyl)purine-2,6-dione
Molekylvægt (g/mol) 186.2
Formel vægt 186.1024
Procent renhed >95