Få mere at vide
Theobromine-d3 (7-methyl-d3), TRC
Artikelnummer. 30449861
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30449861 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30449861 Leverandør Toronto Research Chemicals Leverandørnr. TRCT3438035MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Theobromine-d3 (7-methyl-d3)
CAS 65566-69-0
Molekylær formel C7 D3 H5 N4 O2
Mængde 5 mg
Synonym Theobromine D3 (7-Methyl D3), 3,7-Dihydro-3-methyl-7-(methyl-d3)-1H-purine-2,6-dione (ACI), 1H-Purine-2,6-dione','3,7-dihydro-3-methyl-7-(methyl-d3)- (9CI','ACI)
InChI formel InChI=1S/C7H8N4O2/c1-10-3-8-5-4(10)6(12)9-7(13)11(5)2/h3H,1-2H3,(H,9,12,13)/i1D3
SMIL [2H]C([2H])([2H])n1cnc2N(C)C(=O)NC(=O)c12
IUPAC navn 3-methyl-7-(trideuteriomethyl)purine-2,6-dione
Molekylvægt (g/mol) 183.183
Formel vægt 183.084