Learn More
Theobromine, 99%, Thermo Scientific Chemicals
Catalog Number p-7052397
Click to view available options
Quantity:
50 g
100 g
250 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
83-67-0
C7H8N4O2
180.167
MFCD00022830
YAPQBXQYLJRXSA-UHFFFAOYSA-N
theobromine, 3,7-dimethylxanthine, diurobromine, teobromin, theosalvose, theostene, santheose, thesodate, thesal, theobromin
5429
CHEBI:28946
3,7-dimethylpurine-2,6-dione
CN1C=NC2=C1C(=O)NC(=O)N2C
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
83-67-0
C7H8N4O2
180.167
MFCD00022830
YAPQBXQYLJRXSA-UHFFFAOYSA-N
theobromine, 3,7-dimethylxanthine, diurobromine, teobromin, theosalvose, theostene, santheose, thesodate, thesal, theobromin
5429
CHEBI:28946
3,7-dimethylpurine-2,6-dione
CN1C=NC2=C1C(=O)NC(=O)N2C
Phosphodiesterase inhibitor
Phosphodiesterase inhibitor, weak adenosine receptor antagonist, diuretic and smooth muscle relaxant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phosphodiesterase inhibitor, weak adenosine receptor antagonist, diuretic and smooth muscle relaxant.
Solubility
Soluble in hot methanol and dimethyl sulfoxide.
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent. Store away from strong bases.
TRUSTED_SUSTAINABILITY
Melting Point 345°C to 350°C
Color White
Density 1.50 g/mL
Quantity 50 g
Beilstein 16464
Merck Index 14,9282
Solubility Information Soluble in hot methanol and dimethyl sulfoxide.
Formula Weight 180.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Theobromine