Få mere at vide
Thenyldiamine Hydrochloride, Mikromol™
Beskrivelse
Thenyldiamine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Thenyldiamine Hydrochloride |
| CAS | 958-93-0 |
| Analyt- eller komponentnavne | Thenyldiamine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H19 N3 S . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | 1,2-Ethanediamine, N1,N1-dimethyl-N2-2-pyridinyl-N2-(3-thienylmethyl)-, hydrochloride (1:1), 1,2-Ethanediamine, N,N-dimethyl-N'-2-pyridinyl-N'-(3-thienylmethyl)-, monohydrochloride (9CI), N,N-Dimethyl-N'-(3-thenyl)-N'-(2-pyridyl)ethylenediamine hydrochloride, Thenfadil hydrochloride, Thenyldiamine hydrochloride |
| InChI formel | InChI=1S/C14H19N3S.ClH/c1-16(2)8-9-17(11-13-6-10-18-12-13)14-5-3-4-7-15-14;/h3-7,10,12H,8-9,11H2,1-2H3;1H |
| SMIL | Cl.CN(C)CCN(Cc1ccsc1)c2ccccn2 |
| IUPAC navn | N,N-dimethyl-N'-pyridin-2-yl-N'-(thiophen-3-ylmethyl)ethane-1,2-diamine;hydrochloride |
| Vis mere |